Learn More
US Plastic Corporation 1/8" ID x 1/4" OD x 1/16" Wall Tygon 2375 Ultra Chemical Resistant Tubing, 10'/pk
SDP
Catalog No. NC1363091
Encompass
This tubing offers an unequaled combination of chemical resistance, clarity and flexibility. It is virtually unaffected by acids, bases, ketones, salts and alcohols, enabling it to meet the requirements of many applications, from filling battery acid to handling hazardous material. With an exceptionally smooth inner bore, the potential for contamination is reduced because particulate buildup is inhibited.
Catalog No. NC1363091 Supplier US Plastic Corporation Supplier No. 5754710FT
This tubing offers an unequaled combination of chemical resistance, clarity and flexibility. It is virtually unaffected by acids, bases, ketones, salts and alcohols, enabling it to meet the requirements of many applications, from filling battery acid to handling hazardous material. With an exceptionally smooth inner bore, the potential for contamination is reduced because particulate buildup is inhibited.