Learn More
US Plastic Corporation TYGON 2375 3/8 X 1/2 (50')
SDP
Catalog No. NC1753393
Encompass
3/8"id x 1/2"od x 1/16" wall Tygon 2375 ultra chemical resistant tubing
Catalog No. NC1753393 Supplier US Plastic Corporation Supplier No. 5755250FT
3/8"id x 1/2"od x 1/16" wall Tygon 2375 ultra chemical resistant tubing