Learn More
Accuflow Systems Tygon Tubing, A-60-F IB, 3/8 in ID, 5/8 in OD
Catalog No. NC2879092
Encompass_Preferred
Tygon A-60-F IB is a reinforced thermoplastic elastomer tubing designed for food and beverage dispensing and peristaltic pump applications. It offers temperature resistance and embedded reinforcement for elevated-pressure lines in processing and dispensing systems.
  • 3/8 in inside diameter, 5/8 in outside diameter, 50 ft length.
  • Wall thickness 1/8 in (0.125 in).
  • Reinforced construction with embedded braid for elevated pressure handling.
  • Thermoplastic elastomer formulation, temperature resistant from -75°F to 275°F (-60°C to 135°C).
  • Cream color for visual identification.
  • Suitable for food and beverage dispensing, peristaltic pumps, cosmetic and dairy processing.
Catalog No. NC2879092 Supplier Accuflow Systems Supplier No. APW00029
Tygon A-60-F IB is a reinforced thermoplastic elastomer tubing designed for food and beverage dispensing and peristaltic pump applications. It offers temperature resistance and embedded reinforcement for elevated-pressure lines in processing and dispensing systems.
  • 3/8 in inside diameter, 5/8 in outside diameter, 50 ft length.
  • Wall thickness 1/8 in (0.125 in).
  • Reinforced construction with embedded braid for elevated pressure handling.
  • Thermoplastic elastomer formulation, temperature resistant from -75°F to 275°F (-60°C to 135°C).
  • Cream color for visual identification.
  • Suitable for food and beverage dispensing, peristaltic pumps, cosmetic and dairy processing.