Learn More
Accuflow Systems Saint-Gobain Tygon S3 Tubing, 3/8 in ID, 5/8 in OD
Catalog No. NC2942967
Encompass_Preferred
Tygon S3 B-44-4X is a clear, flexible thermoplastic elastomer tubing designed for sanitary food and beverage dispensing and transfer applications. It features a smooth, non-porous bore for easy cleaning and minimal particle entrapment, and resists alkaline cleaners and sanitizers.
  • Clear, flexible TPE construction for visible flow monitoring.
  • 3/8 in inner diameter, 5/8 in outer diameter, 1/8 in wall thickness.
  • Supplied on 50 ft roll for extended runs.
  • Maximum working pressure approximately 44 psi.
  • Operating temperature range -49°F to 165°F (-45°C to 74°C).
  • Meets common food-contact standards and certifications.
  • Contains no BPA or phthalates; smooth bore reduces bacterial harboring.
Catalog No. NC2942967 Supplier Accuflow Systems Supplier No. AAA00029
Tygon S3 B-44-4X is a clear, flexible thermoplastic elastomer tubing designed for sanitary food and beverage dispensing and transfer applications. It features a smooth, non-porous bore for easy cleaning and minimal particle entrapment, and resists alkaline cleaners and sanitizers.
  • Clear, flexible TPE construction for visible flow monitoring.
  • 3/8 in inner diameter, 5/8 in outer diameter, 1/8 in wall thickness.
  • Supplied on 50 ft roll for extended runs.
  • Maximum working pressure approximately 44 psi.
  • Operating temperature range -49°F to 165°F (-45°C to 74°C).
  • Meets common food-contact standards and certifications.
  • Contains no BPA or phthalates; smooth bore reduces bacterial harboring.