Learn More
Braintree Scientific Tygon Catheter Tubing, 50 ft. coil, .06""""OD x .02""""ID
SDP
Catalog No. 501956085
Encompass
Tygon Catheter TubingFlexible and clear; this tubing is ideal for cannulations. It is easily fabricated to various catheter styles and is virtually kink free when securing in a vessel and exiting the animal model. This material is less permeable than silicon rubber tubing and has a very smooth surface which inhibits thrombic events. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6085 Supplier Braintree Scientific Supplier No. TYG060
Tygon Catheter TubingFlexible and clear; this tubing is ideal for cannulations. It is easily fabricated to various catheter styles and is virtually kink free when securing in a vessel and exiting the animal model. This material is less permeable than silicon rubber tubing and has a very smooth surface which inhibits thrombic events. **Sold for research purposes only and not for human use**