Learn More
Accuflow Systems Saint-Gobain Tygon E-1000 Tubing, 0.25 in ID, 50 ft
Catalog No. NC2958511
Encompass_Preferred
Tygon E-1000 ultra-soft laboratory tubing delivers low-durometer flexibility for peristaltic pump and laboratory dispensing applications that require tight bends and low torque. The thermoplastic elastomer construction maintains flexibility at low temperatures and is formulated with non-DEHP plasticizers. This configuration is 0.250 in inner diameter, 0.375 in outer diameter, and supplied in 50 ft lengths.
  • Thermoplastic elastomer material, 40 Shore A durometer.
  • 0.250 in inner diameter, 0.375 in outer diameter, 50 ft length.
  • Temperature range -67°F to 125°F (-55°C to 52°C).
  • Formulated with non-DEHP plasticizers.
  • Suitable for peristaltic pumps and low-torque pumping.
  • Resists twisting and collapse; maintains flexibility at low temperatures.
Catalog No. NC2958511 Supplier Accuflow Systems Supplier No. ADK00017
Tygon E-1000 ultra-soft laboratory tubing delivers low-durometer flexibility for peristaltic pump and laboratory dispensing applications that require tight bends and low torque. The thermoplastic elastomer construction maintains flexibility at low temperatures and is formulated with non-DEHP plasticizers. This configuration is 0.250 in inner diameter, 0.375 in outer diameter, and supplied in 50 ft lengths.
  • Thermoplastic elastomer material, 40 Shore A durometer.
  • 0.250 in inner diameter, 0.375 in outer diameter, 50 ft length.
  • Temperature range -67°F to 125°F (-55°C to 52°C).
  • Formulated with non-DEHP plasticizers.
  • Suitable for peristaltic pumps and low-torque pumping.
  • Resists twisting and collapse; maintains flexibility at low temperatures.