Learn More
Component Supply Tygon E-3603 Laboratory Tubing, 1/8'' I.D. 3/16'' O.D.
SDP
Catalog No. NC1807906
Encompass
This item has a minimum qty of 5 per supplier requirements.
Tygon E-3603 offers the superior performance traditionally found in Tygon products while addressing the increasing safety concerns regarding phthalates and BPA as well as the increased regulation of products using plasticizers containing DEHP.
Catalog No. NC1807906 Supplier Component Supply Supplier No. TET125A
This item has a minimum qty of 5 per supplier requirements.
Tygon E-3603 offers the superior performance traditionally found in Tygon products while addressing the increasing safety concerns regarding phthalates and BPA as well as the increased regulation of products using plasticizers containing DEHP.