Learn More
AVANTOR MASTERFLEX (VWR LLC) Tygon E-LFL Pump Tubing, B/T 91, 10'
Catalog No. NC0694193
Encompass
Masterflex B/T Tygon E-LFL Pump Tubing, B/T 91, 10 ft. High-performance pump tubing for challenging applicationsNontoxic and nonhemolytic with excellent biocompatibilityLong pumping life; pump at continuous pressures up to 40 psiLow extractables and low gas permeabilityMeets USP Class VI criteria and is FDA compliantB/T 91 tubing has 3/4¿ (19 mm) inner diameter
Catalog No. NC0694193 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0644091/DEL
Masterflex B/T Tygon E-LFL Pump Tubing, B/T 91, 10 ft. High-performance pump tubing for challenging applicationsNontoxic and nonhemolytic with excellent biocompatibilityLong pumping life; pump at continuous pressures up to 40 psiLow extractables and low gas permeabilityMeets USP Class VI criteria and is FDA compliantB/T 91 tubing has 3/4¿ (19 mm) inner diameter