Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, S3, 1/4 in ID, 1/2 in OD
Catalog No. 502365883
Encompass_Preferred
Tygon E-3603 (S3 formulation) is a crystal-clear, flexible thermoplastic tubing designed for medical and laboratory fluid transfer applications that require chemical compatibility, visual inspection, and low extractables. Heavy-wall construction provides vacuum integrity and resistance to flex-fatigue and abrasion, making it suitable for peristaltic pump and diagnostic instrument use.
  • Crystal-clear, flexible thermoplastic tubing
  • Heavy wall 1/8 in (3.2 mm) for vacuum integrity
  • Inner diameter 1/4 in (6.4 mm), outer diameter 1/2 in (12.7 mm)
  • 50 ft (15 m) coil length standard
  • Maximum recommended operating temperature 165°F (74°C)
  • BPA- and phthalate-free, non-oxidizing formulation that minimizes extractables
  • Suitable for peristaltic pump and general laboratory fluid transfer applications
Catalog No. 50-236-5883 Supplier Accuflow Systems Supplier No. ACF00019
Tygon E-3603 (S3 formulation) is a crystal-clear, flexible thermoplastic tubing designed for medical and laboratory fluid transfer applications that require chemical compatibility, visual inspection, and low extractables. Heavy-wall construction provides vacuum integrity and resistance to flex-fatigue and abrasion, making it suitable for peristaltic pump and diagnostic instrument use.
  • Crystal-clear, flexible thermoplastic tubing
  • Heavy wall 1/8 in (3.2 mm) for vacuum integrity
  • Inner diameter 1/4 in (6.4 mm), outer diameter 1/2 in (12.7 mm)
  • 50 ft (15 m) coil length standard
  • Maximum recommended operating temperature 165°F (74°C)
  • BPA- and phthalate-free, non-oxidizing formulation that minimizes extractables
  • Suitable for peristaltic pump and general laboratory fluid transfer applications