Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon F-4040-A, Fuel and Lubricant, 1/8"" ID x 1/4"" OD; 50 ft
Catalog No. NC0330817
Encompass
The Tygon tubing is ideal for fuels and industrial lubricants¿gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. Resists most hydrocarbons, embrittlement and swelling, ozone- and UV-resistant, with low-extractability. Tubing is manufactured according to GMP and has a temperature range of -35 to 165¿F (-37 to 74¿C). Sterilization is not recommended.
Catalog No. NC0330817 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9563301
The Tygon tubing is ideal for fuels and industrial lubricants¿gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. Resists most hydrocarbons, embrittlement and swelling, ozone- and UV-resistant, with low-extractability. Tubing is manufactured according to GMP and has a temperature range of -35 to 165¿F (-37 to 74¿C). Sterilization is not recommended.