Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon 2475 High Purity, 1/4"" ID x 3/8"" OD; 50 ft
Catalog No. NC0174124
Encompass
Formulation: TYGON 2475 Applications: Sensitive fluid transfer applications requiring high purity. Characteristics: Plasticizer free, smooth inner surface (inhibits particulate build-up and bacterial growth), safely dispose of through incineration, and nontoxic. Transparent. Certification: Resin is FDA compliant and meets USP 23 Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -94 to 130¿F (-70 to 54¿C) Sterilize: With ethylene oxide or gamma irradiation.
Catalog No. NC0174124 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9563553
Formulation: TYGON 2475 Applications: Sensitive fluid transfer applications requiring high purity. Characteristics: Plasticizer free, smooth inner surface (inhibits particulate build-up and bacterial growth), safely dispose of through incineration, and nontoxic. Transparent. Certification: Resin is FDA compliant and meets USP 23 Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -94 to 130¿F (-70 to 54¿C) Sterilize: With ethylene oxide or gamma irradiation.