Learn More
AVANTOR MASTERFLEX (VWR LLC) Tygon ACF00001 E-3603 Non-DEHP Nominal Lab Tubing, 1/32"" ID x 3/32"" OD; 50 ft/Pk
Catalog No. NC0894938
Encompass
Formulation - E3603 Applications:General laboratory use. Characteristics: Long life, won¿t age or oxidize, able to bend to sharp radius curves, and easily works with fittings. Transparent. Temperature range: ¿58 to 165¿F (¿50 to 74¿C)
Catalog No. NC0894938 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0740770
Formulation - E3603 Applications:General laboratory use. Characteristics: Long life, won¿t age or oxidize, able to bend to sharp radius curves, and easily works with fittings. Transparent. Temperature range: ¿58 to 165¿F (¿50 to 74¿C)