Learn More
AVANTOR MASTERFLEX (VWR LLC) E-3603 Non-DEHP Lab Tubing, 1/8"" ID x 1/4"" OD; 50 ft/Pk
Catalog No. NC0467390
Encompass
E-3603 Non-DEHP Lab Tubing, 1/8" ID x 1/4" OD; 50 ft/PkFormulation: Tygon E-3603Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments.Characteristics: Non-DEHP, phthalate free. Long life, chemical resistant, nonoxidizing; able to bend to sharp radius curves and easily works with fittings. Transparent.Certifications: FDA compliant; meets USP Class VI and NSF 51 requirements.Temperature range: ¿51 to 165¿F (¿46 to 74¿C)Sterilize: Autoclave or ethylene oxide.
Catalog No. NC0467390 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640776
E-3603 Non-DEHP Lab Tubing, 1/8" ID x 1/4" OD; 50 ft/PkFormulation: Tygon E-3603Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments.Characteristics: Non-DEHP, phthalate free. Long life, chemical resistant, nonoxidizing; able to bend to sharp radius curves and easily works with fittings. Transparent.Certifications: FDA compliant; meets USP Class VI and NSF 51 requirements.Temperature range: ¿51 to 165¿F (¿46 to 74¿C)Sterilize: Autoclave or ethylene oxide.