Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon ND-100-80 Microbore, 0.010"" ID x 0.030"" OD; 100 ft
Catalog No. NC0597887
Encompass
Formulation: Tygon ND-100-80 Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications. Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear. Certifications: Resin meets USP Class VI requirements. Temperature range: -25 to 185¿F (-31 to 85¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
Catalog No. NC0597887 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641900
Formulation: Tygon ND-100-80 Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications. Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear. Certifications: Resin meets USP Class VI requirements. Temperature range: -25 to 185¿F (-31 to 85¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.