Learn More
Accuflow Systems Saint-Gobain Tygon ND Tubing, 3/8 in ID, 5/8 in OD
Supplier:  Accuflow Systems ADF00029
Encompass_Preferred
Tygon ND 100-65 tubing is a clear, medical-grade, non-DEHP tubing engineered for regulated fluid transfer and blood-contact applications. It offers robust biocompatibility, easy bonding and welding, and compatibility with common sterilization methods.
  • Clear for visual inspection of fluid flow.
  • 3/8 in inside diameter, 5/8 in outside diameter, 1/8 in wall thickness.
  • Available in 50 ft lengths.
  • Non-DEHP formulation.
  • Tested to elements of ISO 10993 and meets USP <88> Class VI requirements.
  • Compatible with steam, gamma, and ethylene oxide sterilization.
  • Maximum recommended operating temperature 165°F (74°C).
  • Brittleness by impact temperature -44°F (-42°C).
  • Bondable and weldable by heat, RF/ultrasonic, solvent, or adhesive methods.
  • Cleanroom manufacturing available.
Catalog No. 50-236-5870