Learn More
AVANTOR MASTERFLEX (VWR LLC) Tubing, 3-Stop LMT-55, 0.19mm ID, 12/pk
Catalog No. NC0456571
Encompass
Ismatec SC0189T 3-Stop LMT-55 Tubing, 0.19mm ID, 12/pk. Designed for optimal operation in your Ismatec pumpsThe overall length of LMT-55 97619-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Catalog No. NC0456571 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9761909
Ismatec SC0189T 3-Stop LMT-55 Tubing, 0.19mm ID, 12/pk. Designed for optimal operation in your Ismatec pumpsThe overall length of LMT-55 97619-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.