Learn More
Perkin Elmer US LLC Tygon Rinse/Drain Tubing Hookup Kit for XLR-860, Aqueous Applications
Catalog No. 502104706
Encompass_Preferred
It includes: 1/8 X 3/16 PVDF (KYNAR) couplers (qty. 2) 22.5 (0.6 m) of 3/16 (4.8 mm) I.D. Tygon tubing 256 (6.5 m) of 1/8 (3.2 mm) I.D. Tygon tubing
Catalog No. 50-210-4706 Supplier Perkin Elmer US LLC Supplier No. N0782077
It includes: 1/8 X 3/16 PVDF (KYNAR) couplers (qty. 2) 22.5 (0.6 m) of 3/16 (4.8 mm) I.D. Tygon tubing 256 (6.5 m) of 1/8 (3.2 mm) I.D. Tygon tubing