Learn More
Accuflow Systems Tygon S3 B-44-4X Tubing, 1 in. ID, 50 ft.
Catalog No. NC3997532
Encompass_Preferred
Tygon® B-44-4X tubing is a lightweight, flexible, and clear tubing designed for reliable performance in food, milk, and dairy processing applications such as aseptic filling, condiment dispensing, and transfer. Its smooth, non-porous bore inhibits particle entrapment and bacterial growth, ensuring a sanitary fluid path.
  • Smooth, non-porous bore inhibits particle entrapment and bacterial growth.
  • Resistant to harsh alkaline cleaners and sanitizers.
  • Excellent alternative to rigid piping systems.
  • Contains no BPA or phthalates.
  • Compatible with high oil content foods.
  • Glass-like clarity allows visual monitoring of product flow.
Catalog No. NC3997532 Supplier Accuflow Systems Supplier No. AAA00062
Tygon® B-44-4X tubing is a lightweight, flexible, and clear tubing designed for reliable performance in food, milk, and dairy processing applications such as aseptic filling, condiment dispensing, and transfer. Its smooth, non-porous bore inhibits particle entrapment and bacterial growth, ensuring a sanitary fluid path.
  • Smooth, non-porous bore inhibits particle entrapment and bacterial growth.
  • Resistant to harsh alkaline cleaners and sanitizers.
  • Excellent alternative to rigid piping systems.
  • Contains no BPA or phthalates.
  • Compatible with high oil content foods.
  • Glass-like clarity allows visual monitoring of product flow.