Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, BioPharm Plus Platinum-Cured Silicone, L/S 17; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9644017/DEL
Encompass
Applications: Pharmaceutical, biotechnology, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Choose our BioPharm Plus Silicone (Platinum-Cured) tubing for purity, flexibility, and longer tubing life. Ideal for pharmaceutical and biotechnological applications, this tubing exceeds USP class VI, 3A sanitary standards, and is FDA compliant. Based on your application needs, choose Precision tubing or High-Performance precision tubing. High-Performance precision tubing has a thicker wall that improves pressure generation suction lift and the ability to pump viscous fluids. BioPharm Plus Silicone (Platinum-Cured) tubing has a working temperature range of ¿75 to 450¿F (¿60 to 232¿C). Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide.
Catalog No. NC9587401