Learn More
AVANTOR MASTERFLEX (VWR LLC) Lab Tubing, Non-DEHP, 1/8""ID x 1/4""OD, 50 ft/pack
Catalog No. NC0603997
Encompass
Tygon ACF00007 E-3603 Non-DEHP Nominal Lab Tubing, 1/8" ID x 1/4" OD; 50 ft/PkLong life, won¿t age or oxidize. Formulation - E3603Applications:General laboratory use.Characteristics: Long life, won¿t age or oxidize, able to bend to sharp radius curves, and easily works with fittings. Transparent.Temperature range: ¿58 to 165¿F (¿50 to 74¿C)
Catalog No. NC0603997 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0740776
Tygon ACF00007 E-3603 Non-DEHP Nominal Lab Tubing, 1/8" ID x 1/4" OD; 50 ft/PkLong life, won¿t age or oxidize. Formulation - E3603Applications:General laboratory use.Characteristics: Long life, won¿t age or oxidize, able to bend to sharp radius curves, and easily works with fittings. Transparent.Temperature range: ¿58 to 165¿F (¿50 to 74¿C)