Learn More
Sigma Aldrich Fine Chemicals Biosciences Tygon formula E-3603 laboratory tubing, 50ft
Catalog No. NC1598624
Encompass
I.D. × O.D. × wall thickness: 3/16 in. × 9/16 in. × 3/16 in. TYGON E-3603 non-DEHP laboratory and vacuum tubing delivers the same superior performance you′ve come to expect from TYGON products - but now in a formulation that contains a widely accepted substitute for DEHP plasticizer. TYGON E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion, handles virtually all inorganic chemicals found in the lab, and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.
Catalog No. NC1598624 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z76531750FT
I.D. × O.D. × wall thickness: 3/16 in. × 9/16 in. × 3/16 in. TYGON E-3603 non-DEHP laboratory and vacuum tubing delivers the same superior performance you′ve come to expect from TYGON products - but now in a formulation that contains a widely accepted substitute for DEHP plasticizer. TYGON E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion, handles virtually all inorganic chemicals found in the lab, and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.