Learn More
Grainger TYGON Tubing, 1/32" - 3/32", 50FT
Catalog No. NC2883460
Encompass_Preferred
The Tygon Tubing 1/32" ID, 3/32" OD, 50' Length is flexible PVC tubing that is clear and smooth. It has a 1/32" inner diameter and 3/32" outer diameter. The tubing is 50 feet in length and is sold by the foot. It has a maximum pressure rating of 30 psi at 73°F. The tubing is non-toxic, tasteless, and odorless. It is flexible and kink resistant. The tubing has applications for transferring fluids and gases, instrument connections, aquariums, and more. The tubing meets FDA, USP Class VI, and UL standards. It has a temperature range of -60°F to 160°F.
Catalog No. NC2883460 Supplier Grainger Supplier No. 22XH28
The Tygon Tubing 1/32" ID, 3/32" OD, 50' Length is flexible PVC tubing that is clear and smooth. It has a 1/32" inner diameter and 3/32" outer diameter. The tubing is 50 feet in length and is sold by the foot. It has a maximum pressure rating of 30 psi at 73°F. The tubing is non-toxic, tasteless, and odorless. It is flexible and kink resistant. The tubing has applications for transferring fluids and gases, instrument connections, aquariums, and more. The tubing meets FDA, USP Class VI, and UL standards. It has a temperature range of -60°F to 160°F.