Learn More
Glass Expansion Tygon Tubing 1/4 inch x 1m
SDP
Catalog No. NC0015018
Encompass_Preferred
Tygon Tubing 1/4 inch x 1m. A 1 meter length of 1/4 inch Tygon tubing. Compatible with GAZ-08. An example of use is as a soft walled transfer tube between Glass Expansion’s IsoMist (e.g., KT-1064-XR) spray chamber and ICP torch using GazFit Connector 8mm (GAZ-08) and a Glass Ball Cup Adaptor FS13 (70-803-0571).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC0015018 Supplier Glass Expansion Supplier No. 19211M
Tygon Tubing 1/4 inch x 1m. A 1 meter length of 1/4 inch Tygon tubing. Compatible with GAZ-08. An example of use is as a soft walled transfer tube between Glass Expansion’s IsoMist (e.g., KT-1064-XR) spray chamber and ICP torch using GazFit Connector 8mm (GAZ-08) and a Glass Ball Cup Adaptor FS13 (70-803-0571).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001