Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon 2375, 3/8"" ID x 1/2"" OD; 50 ft
Catalog No. NC0378794
Encompass
These transparent tubes are ideal for use with highly aggressive chemicals ¿ acids, bases, ketones, salts, and alcohols. Characteristics include plasticizer free, smoother inner surface, hydrophobic, minimal outgassing, safe disposal via incineration, and nontoxic. NSF 51 Standard compliant and manufactured according to GMP. Temperature range is -108 to 130¿F (-78 to 54¿C). Can be sterilized with ethylene oxide or gamma irradiation.
Catalog No. NC0378794 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9563018
These transparent tubes are ideal for use with highly aggressive chemicals ¿ acids, bases, ketones, salts, and alcohols. Characteristics include plasticizer free, smoother inner surface, hydrophobic, minimal outgassing, safe disposal via incineration, and nontoxic. NSF 51 Standard compliant and manufactured according to GMP. Temperature range is -108 to 130¿F (-78 to 54¿C). Can be sterilized with ethylene oxide or gamma irradiation.