Learn More
AVANTOR MASTERFLEX (VWR LLC) TYGON TUBING E3603 3/32X7/32
Catalog No. NC2003102
Encompass
Masterflex Transfer Tubing, Tygon S3 E-3603, 3/32" ID x 7/32" OD; 50 ft
Catalog No. NC2003102 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640774
Masterflex Transfer Tubing, Tygon S3 E-3603, 3/32" ID x 7/32" OD; 50 ft