Learn More
Accuflow Systems Saint-Gobain Tygon Power Tubing, 1/2 in ID, 50 ft
Catalog No. NC2478549
Encompass_Preferred
Tygon Power F-4040-A tubing is a hydrocarbon-resistant PVC formulation designed for fuel and lubricant transfer. It resists swelling and hardening from hydrocarbon-based fluids, offers low extractables to reduce contamination risk, and is suitable for small-engine fuel lines, automotive, and industrial lubrication applications.
  • Hydrocarbon-resistant PVC formulation for fuel and lubricant service.
  • Inner diameter 1/2 in, outer diameter 3/4 in, wall thickness 1/8 in.
  • Standard length 50 ft roll.
  • Yellow color for fluid identification.
  • Hardness 57 Shore A.
  • Maximum recommended operating temperature 165°F (74°C).
  • Low extractables to minimize contamination of transferred fluids.
  • Typical uses: small engine fuel lines, general automotive, recreational vehicles, lawn and garden equipment, coolant transfer, heating fuels, cutting compounds, polishing equipment, lubrication lines.
Catalog No. NC2478549 Supplier Accuflow Systems Supplier No. AAG00038
Tygon Power F-4040-A tubing is a hydrocarbon-resistant PVC formulation designed for fuel and lubricant transfer. It resists swelling and hardening from hydrocarbon-based fluids, offers low extractables to reduce contamination risk, and is suitable for small-engine fuel lines, automotive, and industrial lubrication applications.
  • Hydrocarbon-resistant PVC formulation for fuel and lubricant service.
  • Inner diameter 1/2 in, outer diameter 3/4 in, wall thickness 1/8 in.
  • Standard length 50 ft roll.
  • Yellow color for fluid identification.
  • Hardness 57 Shore A.
  • Maximum recommended operating temperature 165°F (74°C).
  • Low extractables to minimize contamination of transferred fluids.
  • Typical uses: small engine fuel lines, general automotive, recreational vehicles, lawn and garden equipment, coolant transfer, heating fuels, cutting compounds, polishing equipment, lubrication lines.