Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, Tygon Chemical Tubing, L/S 25; 50 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0647525/DEL
Encompass
Applications: Chemical, food, beverage, and pharmaceutical applications Characteristics: Excellent chemical resistance, low extractability, low gas permeability, plasticizer free, translucent NOTE: Tygon Chemical pump tubing is recommended for use in Easy-Load and Easy-Load II pump heads only.
Catalog No. NC0435398
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.