Learn More
AVANTOR MASTERFLEX (VWR LLC) Tygon ACF00028 E-3603 Non-DEHP Nominal Lab Tubing, 3/8"" ID x 9/16"" OD; 50 ft/Pk
Catalog No. NC1596480
Encompass
Formulation - E3603 Applications:General laboratory use. Characteristics: Long life, won¿t age or oxidize, able to bend to sharp radius curves, and easily works with fittings. Transparent. Temperature range: ¿58 to 165¿F (¿50 to 74¿C)
Catalog No. NC1596480 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0740787
Formulation - E3603 Applications:General laboratory use. Characteristics: Long life, won¿t age or oxidize, able to bend to sharp radius curves, and easily works with fittings. Transparent. Temperature range: ¿58 to 165¿F (¿50 to 74¿C)