Tygon S3 E-3603 laboratory tubing is a clear, flexible thermoplastic elastomer tubing designed for reliable laboratory fluid handling. It provides a glassy smooth inner bore for easy cleaning, broad compatibility with organic chemicals, and resistance to flex-fatigue and abrasion for extended service life. The tubing is suitable for peristaltic pumps, instrumentation connections, vents, drains, and general laboratory transfer applications.
- Thermoplastic elastomer construction for chemical resistance and flexibility.
- Inner diameter 2.38 mm (3/32 in), outer diameter 5.56 mm (7/32 in), wall thickness 1.59 mm (1/16 in).
- Temperature range -46°C to 74°C (-51°F to 165°F).
- Full-vacuum compatible at room temperature; working pressure varies by size.
- Handles most organic laboratory chemicals.
- Phthalate-free, non-DEHP formulation; contains no BPA.
- Resistant to flex-fatigue and abrasion for extended service life.