Learn More
Sigma Aldrich Fine Chemicals Biosciences TygonR formula 2375 labora
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6855851PAK
Encompass
Tygon laboratory tubing formula 2375 is chemically resistant tubing. Clear flexible tubing suitable for specialty chemical production and hazardous material handling.Resistant to MEK acids bases ketones and alcoholsPlasticizer freeNo contamination from leachingBiocompatibleNo disposal concernsReleases only carbon dioxide and water upon incinerationSterilize by gas or radiation
Catalog No. 50-290-1451