Learn More
Sigma Aldrich Fine Chemicals Biosciences TygonR formula E-3603 labo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z76524450FT
Encompass
TYGON(R) E-3603 non-DEHP laboratory and vacuum tubing delivers the same superior performance youve come to expect from TYGON products - but now in a formulation that contains a widely accepted substitute for DEHP plasticizer.TYGON(R) E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion handles virtually all inorganic chemicals found in the lab and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.TYGON(R) E-3603 vacuum tubing shares all these performance features and has extra heavy walls that will withstand a full vacuum at room temperature.features/BenefitsAlternative to DEHP tubingNew formulation delivers the same excellent performanceContains no BPAOutstanding chemical resistanceLot-to-lot consistency for reproducible resultsIncreases productivity in peristaltic pumps - outlasts other clear tubing 2 to 1Smooth polished inner boreNon-oxidi
Catalog No. 50-290-5884