Learn More
Accuflow Systems Saint-Gobain Pump Tubing, Tygon XL-60, 1/8 in ID, TPE
Catalog No. 502365842
Encompass_Preferred
Saint-Gobain Tygon XL-60 tubing is a long-life thermoplastic elastomer (TPE) formulated for peristaltic pump applications. It offers extended pump life compared with standard silicone or PVC tubing, remains flexible at low temperatures, and resists elevated operating temperatures.
  • Material: thermoplastic elastomer (TPE), Shore A 60.
  • Designed for peristaltic pump applications, long-life formulation.
  • Internal diameter 1/8 in, outer diameter 1/4 in, wall thickness 1/16 in.
  • Supplied in 50 ft lengths.
  • Temperature range -40°F to 250°F (maximum recommended 250°F / 121°C).
  • Pump life exceeding 500 hours under typical operating conditions.
Catalog No. 50-236-5842 Supplier Accuflow Systems Supplier No. AN800007
Saint-Gobain Tygon XL-60 tubing is a long-life thermoplastic elastomer (TPE) formulated for peristaltic pump applications. It offers extended pump life compared with standard silicone or PVC tubing, remains flexible at low temperatures, and resists elevated operating temperatures.
  • Material: thermoplastic elastomer (TPE), Shore A 60.
  • Designed for peristaltic pump applications, long-life formulation.
  • Internal diameter 1/8 in, outer diameter 1/4 in, wall thickness 1/16 in.
  • Supplied in 50 ft lengths.
  • Temperature range -40°F to 250°F (maximum recommended 250°F / 121°C).
  • Pump life exceeding 500 hours under typical operating conditions.