Learn More
Accuflow Systems Saint-Gobain Versilon Tubing, Ester-based Polyurethane, 1/4 in ID, 100 ft
Catalog No. 502365806
Encompass_Preferred
Versilon C-210-A (Tygothane C-210-A) is a clear, ester-based polyurethane tubing designed for chemical transfer, fuel and lubricant lines, and pneumatic systems. Plasticizer-free construction provides flexibility across extreme temperatures while offering high tear strength and abrasion resistance for demanding fluid-handling and instrumentation applications.
  • Ester-based polyurethane offers high tear strength and abrasion resistance.
  • Plasticizer-free formulation maintains flexibility in extreme temperatures.
  • Wide operating temperature range: -100°F to 175°F (-73°C to 79°C).
  • Available size: 1/4 in ID, 3/8 in OD, 1/16 in wall, 100 ft length.
  • High tensile strength and 500% elongation support rugged use and repeated flexing.
  • Suitable for fuel, lubricant, pneumatic, and abrasive slurry transfer applications.
  • Clear tubing permits visual flow monitoring and contamination detection.
Catalog No. 50-236-5806 Supplier Accuflow Systems Supplier No. AEM02017
Versilon C-210-A (Tygothane C-210-A) is a clear, ester-based polyurethane tubing designed for chemical transfer, fuel and lubricant lines, and pneumatic systems. Plasticizer-free construction provides flexibility across extreme temperatures while offering high tear strength and abrasion resistance for demanding fluid-handling and instrumentation applications.
  • Ester-based polyurethane offers high tear strength and abrasion resistance.
  • Plasticizer-free formulation maintains flexibility in extreme temperatures.
  • Wide operating temperature range: -100°F to 175°F (-73°C to 79°C).
  • Available size: 1/4 in ID, 3/8 in OD, 1/16 in wall, 100 ft length.
  • High tensile strength and 500% elongation support rugged use and repeated flexing.
  • Suitable for fuel, lubricant, pneumatic, and abrasive slurry transfer applications.
  • Clear tubing permits visual flow monitoring and contamination detection.