Learn More
Carnabio Usa Inc TYK2/5UG/CATALYTIC
Catalog No. 507540654
Encompass
TYK2, Catalytic domain, Wild type, Amino Acid 871-1187, NP_003322.3 , Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0654 Supplier Carnabio Usa Inc Supplier No. 081475
TYK2, Catalytic domain, Wild type, Amino Acid 871-1187, NP_003322.3 , Expressed in Insect (sf21); 5microg