Learn More
Medchemexpress LLC HY-111745 10mg Medchemexpress, Tyk2-IN-5 CAS:1797432-62-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11174510MG
Encompass_Preferred
Medchemexpress, HY-111745 10mg Tyk2-IN-5 CAS:1797432-62-2 Tyk2-IN-5 (compound 6) is a highly potent, selective and orally active Tyk2 inhibitor and targets the JH2 domain, with a Ki of 0.086 nM for Tyk2 JH2 and an IC50 of 25 nM for IFNα. Highly effective in inhibiting IFNγ production in a rat pharmacodynamics model and fully efficacious in a rat adjuvant arthritis model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8945