Learn More
Cayman Chemical TYK2 JH2 DomaInhumnrecom 20ug
SDP
Catalog No. 502943283
Encompass_Preferred
Source: Active recombinant human N-terminal DYKDDDDK-tagged TYK2 JH2 domain expressed in insect cells • Amino acids: 575-869 • MW: 34 kDa
Catalog No. 50-294-3283 Supplier Cayman Chemical Supplier No. 4227020 UG
Source: Active recombinant human N-terminal DYKDDDDK-tagged TYK2 JH2 domain expressed in insect cells • Amino acids: 575-869 • MW: 34 kDa