Learn More
Medchemexpress LLC Tylosin | 1401-69-0 | 98.0% | 50 MG
SDP
Catalog No. 501932917
Encompass_Preferred
Tylosin is a macrolide antibiotic naturally produced by Streptomyces fradiae. It exhibits potent antimicrobial activity against Gram-positive bacteria and functions by binding to the 23S rRNA of the bacterial ribosomal 50S subunit, preventing protein synthesis.
  • Effectively combats Gram-positive bacteria
  • Used as a feed additive to promote animal growth
  • Applied in veterinary medicine to treat bacterial dysentery and respiratory diseases in poultry, pigs, and cattle
  • Inhibits protein synthesis by binding to bacterial ribosomal 50S subunit
  • Reduces elevated TNF-α and IL-1β levels and increases IL-10 levels in LPS-treated mice
Catalog No. 50-193-2917 Supplier Medchemexpress LLC Supplier No. HYB0519A50MG
Tylosin is a macrolide antibiotic naturally produced by Streptomyces fradiae. It exhibits potent antimicrobial activity against Gram-positive bacteria and functions by binding to the 23S rRNA of the bacterial ribosomal 50S subunit, preventing protein synthesis.
  • Effectively combats Gram-positive bacteria
  • Used as a feed additive to promote animal growth
  • Applied in veterinary medicine to treat bacterial dysentery and respiratory diseases in poultry, pigs, and cattle
  • Inhibits protein synthesis by binding to bacterial ribosomal 50S subunit
  • Reduces elevated TNF-α and IL-1β levels and increases IL-10 levels in LPS-treated mice