Learn More
Apexbio Technology LLC Tylosin phosphate 1405-53-4 5g
SDP
Catalog No. 5000091180
Encompass_Preferred
Tylosin phosphate (CAS 1405-53-4) is a small-molecule inhibitor targeting bacterial ribosomal subunits It is designed to interfere with bacterial translation thereby inhibiting bacterial protein synthesis Tylosin phosphate exerts its biological activity primarily through binding to ribosomal subunits of Gram-positive bacteria In in vitro studies tylosin phosphate demonstrates inhibitory activity with reported IC50 values generally ranging between 0 5 5 g/mL depending on bacterial strain and assay conditions Based on these pharmacological properties tylosin phosphate holds research potential in studies of antibiotic resistance mechanisms antibacterial spectrum evaluation and as an antibiotic additive to explore growth promotion and therapeutic utility in poultry swine and bovine models
Catalog No. 50-000-91180 Supplier Apexbio Technology LLC Supplier No. B48655000
Tylosin phosphate (CAS 1405-53-4) is a small-molecule inhibitor targeting bacterial ribosomal subunits It is designed to interfere with bacterial translation thereby inhibiting bacterial protein synthesis Tylosin phosphate exerts its biological activity primarily through binding to ribosomal subunits of Gram-positive bacteria In in vitro studies tylosin phosphate demonstrates inhibitory activity with reported IC50 values generally ranging between 0 5 5 g/mL depending on bacterial strain and assay conditions Based on these pharmacological properties tylosin phosphate holds research potential in studies of antibiotic resistance mechanisms antibacterial spectrum evaluation and as an antibiotic additive to explore growth promotion and therapeutic utility in poultry swine and bovine models