Learn More
Medchemexpress LLC Tylosin phosphate | 1405-53-4 | 99.3% | 1 G
SDP
Catalog No. 5000407566
Encompass_Preferred
Tylosin phosphate is a macrolide antibiotic naturally produced through the fermentation of Streptomyces fradiae. It exhibits potent antimicrobial activity against Gram-positive bacteria and is widely utilized as a feed additive to promote animal growth. Tylosin phosphate is also employed in veterinary medicine to combat bacterial dysentery and respiratory diseases in poultry, pigs, and cattle. It works by binding to the 23S rRNA of the bacterial ribosomal 50S subunit.
  • Macrolide antibiotic
  • Naturally produced through fermentation of Streptomyces fradiae
  • Exhibits potent antimicrobial activity against Gram-positive bacteria
  • Utilized as a feed additive to promote animal growth
  • Employed in veterinary medicine to combat bacterial dysentery and respiratory diseases
  • Works by binding to the 23S rRNA of the bacterial ribosomal 50S subunit
Catalog No. 50-004-07566 Supplier Medchemexpress LLC Supplier No. HYB0519B1G
Tylosin phosphate is a macrolide antibiotic naturally produced through the fermentation of Streptomyces fradiae. It exhibits potent antimicrobial activity against Gram-positive bacteria and is widely utilized as a feed additive to promote animal growth. Tylosin phosphate is also employed in veterinary medicine to combat bacterial dysentery and respiratory diseases in poultry, pigs, and cattle. It works by binding to the 23S rRNA of the bacterial ribosomal 50S subunit.
  • Macrolide antibiotic
  • Naturally produced through fermentation of Streptomyces fradiae
  • Exhibits potent antimicrobial activity against Gram-positive bacteria
  • Utilized as a feed additive to promote animal growth
  • Employed in veterinary medicine to combat bacterial dysentery and respiratory diseases
  • Works by binding to the 23S rRNA of the bacterial ribosomal 50S subunit