Learn More
Medchemexpress LLC Tylosin (phosphate) | 1405-53-4 | 99.3% | 500 MG
SDP
Catalog No. 5000408807
Encompass_Preferred
Tylosin phosphate is a macrolide antibiotic naturally produced through the fermentation of Streptomyces fradiae. It demonstrates strong antimicrobial activity against Gram-positive bacteria. It is commonly utilized as a feed additive to promote animal growth and is applied in veterinary medicine for treating bacterial dysentery and respiratory diseases in poultry, pigs, and cattle.
  • Strong antimicrobial activity against Gram-positive bacteria.
  • Used as a feed additive to promote animal growth.
  • Applied in veterinary medicine for bacterial dysentery and respiratory diseases in poultry, pigs, and cattle.
  • Naturally produced macrolide antibiotic.
Catalog No. 50-004-08807 Supplier Medchemexpress LLC Supplier No. HYB0519B500MG
Tylosin phosphate is a macrolide antibiotic naturally produced through the fermentation of Streptomyces fradiae. It demonstrates strong antimicrobial activity against Gram-positive bacteria. It is commonly utilized as a feed additive to promote animal growth and is applied in veterinary medicine for treating bacterial dysentery and respiratory diseases in poultry, pigs, and cattle.
  • Strong antimicrobial activity against Gram-positive bacteria.
  • Used as a feed additive to promote animal growth.
  • Applied in veterinary medicine for bacterial dysentery and respiratory diseases in poultry, pigs, and cattle.
  • Naturally produced macrolide antibiotic.