Learn More
Apexbio Technology LLC Tylosin tartrate 74610-55-2 50mg
SDP
Catalog No. 502240398
Encompass_Preferred
Tylosin tartrate (CAS 74610-55-2) is a small-molecule inhibitor targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting peptide elongation during translation Tylosin tartrate exerts its biological activity primarily through reversible binding to the 50S ribosomal subunit In microbiological assays tylosin tartrate demonstrates antibacterial activity with IC50 values typically ranging from 0 01 to 0 1 g/mL against mycoplasma strains depending on strain sensitivity Based on these pharmacological properties tylosin tartrate holds research potential in the investigation of antimicrobial mechanisms resistance profiles and bacterial physiology associated with poultry and livestock infections
This item is not returnable. View return policy
Catalog No. 50-224-0398 Supplier Apexbio Technology LLC Supplier No. B205950
Tylosin tartrate (CAS 74610-55-2) is a small-molecule inhibitor targeting the bacterial 50S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting peptide elongation during translation Tylosin tartrate exerts its biological activity primarily through reversible binding to the 50S ribosomal subunit In microbiological assays tylosin tartrate demonstrates antibacterial activity with IC50 values typically ranging from 0 01 to 0 1 g/mL against mycoplasma strains depending on strain sensitivity Based on these pharmacological properties tylosin tartrate holds research potential in the investigation of antimicrobial mechanisms resistance profiles and bacterial physiology associated with poultry and livestock infections