Learn More
Medchemexpress LLC Tylosin tartrate | 74610-55-2 | 98.0% | 5 G
SDP
Catalog No. 5000407601
Encompass_Preferred
Tylosin tartrate is a macrolide antibiotic naturally derived from the fermentation of *Streptomyces fradiae*. It exhibits potent antimicrobial activity against Gram-positive bacteria and is commonly used as a feed additive to promote animal growth. Additionally, it is employed in veterinary medicine to treat bacterial dysentery and respiratory diseases in poultry, pigs, and cattle. Tylosin tartrate works by binding to the 23S rRNA of the bacterial ribosomal 50S subunit, thereby exerting its antibacterial effects. It can also prevent the growth of certain Gram-negative strains.
  • Exerts antibacterial effects by binding to 23S rRNA of the bacterial ribosomal 50S subunit.
  • Prevents growth of Gram-negative strains.
  • Generally suppresses elevated TNF-α and IL-1β levels and increases IL-10 levels in Lipopolysaccharide (LPS)-treated animals.
Catalog No. 50-004-07601 Supplier Medchemexpress LLC Supplier No. HYB05195G
Tylosin tartrate is a macrolide antibiotic naturally derived from the fermentation of *Streptomyces fradiae*. It exhibits potent antimicrobial activity against Gram-positive bacteria and is commonly used as a feed additive to promote animal growth. Additionally, it is employed in veterinary medicine to treat bacterial dysentery and respiratory diseases in poultry, pigs, and cattle. Tylosin tartrate works by binding to the 23S rRNA of the bacterial ribosomal 50S subunit, thereby exerting its antibacterial effects. It can also prevent the growth of certain Gram-negative strains.
  • Exerts antibacterial effects by binding to 23S rRNA of the bacterial ribosomal 50S subunit.
  • Prevents growth of Gram-negative strains.
  • Generally suppresses elevated TNF-α and IL-1β levels and increases IL-10 levels in Lipopolysaccharide (LPS)-treated animals.