Learn More
Medchemexpress LLC Tylosin tartrate | 74610-55-2 | MFCD07783824 | 98.0% | 100 MG
SDP
Catalog No. 5000408805
Encompass_Preferred
Tylosin tartrate, a macrolide antibiotic from Streptomyces fradiae, shows potent antimicrobial activity against Gram-positive bacteria. It's used as a feed additive for animal growth and in veterinary medicine for bacterial dysentery and respiratory diseases in poultry, pigs, and cattle. Its antibacterial action involves binding to the 23S rRNA of the bacterial ribosomal 50S subunit, and it also inhibits Gram-negative strains. Studies show it modulates TNF-α, IL-1β, and IL-10 levels in LPS-treated animal models.
  • Potent antimicrobial activity against Gram-positive bacteria
  • Acts as a feed additive for animal growth promotion
  • Combats bacterial dysentery and respiratory diseases in livestock
  • Binds to the 23S rRNA of bacterial ribosomal 50S subunit
  • Inhibits growth of Gram-negative strains
  • Modulates TNF-α, IL-1β, and IL-10 levels in LPS-treated animal models
Catalog No. 50-004-08805 Supplier Medchemexpress LLC Supplier No. HYB0519100MG
Tylosin tartrate, a macrolide antibiotic from Streptomyces fradiae, shows potent antimicrobial activity against Gram-positive bacteria. It's used as a feed additive for animal growth and in veterinary medicine for bacterial dysentery and respiratory diseases in poultry, pigs, and cattle. Its antibacterial action involves binding to the 23S rRNA of the bacterial ribosomal 50S subunit, and it also inhibits Gram-negative strains. Studies show it modulates TNF-α, IL-1β, and IL-10 levels in LPS-treated animal models.
  • Potent antimicrobial activity against Gram-positive bacteria
  • Acts as a feed additive for animal growth promotion
  • Combats bacterial dysentery and respiratory diseases in livestock
  • Binds to the 23S rRNA of bacterial ribosomal 50S subunit
  • Inhibits growth of Gram-negative strains
  • Modulates TNF-α, IL-1β, and IL-10 levels in LPS-treated animal models