Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tylosin tartrate, 94%

Catalog No. aaj6263303
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6263303 1 g
AAJ6263306 5 g
AAJ6263309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6263303 Supplier Thermo Scientific Chemicals Supplier No. J6263303
Only null left
Add to Cart

Macrolide antibiotic | CAS: 1405-54-5 | C50H83NO23 | 1066.198 g/mol

It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-54-5
Molecular Formula C50H83NO23
Molecular Weight (g/mol) 1066.198
MDL Number MFCD00070094
InChI Key ICVKYYINQHWDLM-HYHSIFHGSA-N
Synonym Pharmasin; [R-(R'R')]-2,3-Dihydroxybutanedionate Tylosin salt
PubChem CID 131673832
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;2-[(4R,5S,6S,7R,9R,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimetho
SMILES CCC1C(C=C(C=CC(=O)C(CC(C(C(C(CC(=O)O1)O)C)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)C)COC4C(C(C(C(O4)C)O)OC)OC.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Type Tylosin Tartrate
Physical Form Powder
Color Cream to White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 1066.2
Percent Purity 95%
Chemical Name or Material Tylosin tartrate
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 215-781-5
RTECSNumber YP0875000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.