Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tylosin Tartrate, G-Biosciences
SCI_ED

Catalog No. S139769
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139769 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139769 Supplier G-Biosciences Supplier No. RC1127
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tylosin tartrate is a macrolide antibiotic isolated from Streptomyces fradiae.

It is a mixture of tylosin A, B, C, and D tartrates effective against gram-positive bacteria and Mycoplasma species. It has been used to study protein synthesis.

Chemical Identifiers

Molecular Weight (g/mol) 1066.19 g/mol

Specifications

Quantity 5 g
Percent Purity 95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Chemical Name or Material Tylosin Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.