Learn More
Crescent Chemical Co Inc TYLOSINE-TARTRATE SOLN (100X)
SDP
Catalog No. 50249144
Encompass_Preferred
Tylosine-tartrate solution (100x) sterile filtered Macrocyclic antibiotic with large lactone ring. Inhibits bacterial protein synthesis at the ribosomal 50S-subunit. Acts bacteriostatic, in higher concentrations as well bactericidal. Active against Gram-positive bacteria and mycop
Catalog No. 50-249-144 Supplier Crescent Chemical Co Inc Supplier No. SE47988.01
Tylosine-tartrate solution (100x) sterile filtered Macrocyclic antibiotic with large lactone ring. Inhibits bacterial protein synthesis at the ribosomal 50S-subunit. Acts bacteriostatic, in higher concentrations as well bactericidal. Active against Gram-positive bacteria and mycop