Learn More
Abcam Tylo x apol, 25G
Catalog No. 502671242
Encompass_Preferred
MW 280.4 Da. A 10% solution of the nonionic liquid surfactant Tyloxapol in water. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1242 Supplier Abcam Supplier No. AB28542925G
MW 280.4 Da. A 10% solution of the nonionic liquid surfactant Tyloxapol in water. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement