Learn More
Selleck Chemical LLC Tyloxapol S4578-1g
SDP
Catalog No. 503108941
Encompass_Preferred
Tyloxapol (Triton WR1339) is a nonionic liquid polymer of the alkyl aryl polyether alcohol type used as a surfactant to aid liquefaction removal of mucopurulent and bronchopulmonary secretions It also blocks plasma lipolytic activity and thus the breakdown of triglyceride-rich lipoproteins Tyloxapol can be used to induce animal models of Hyperlipidemia Atherosclerosis
Catalog No. 50-310-8941 Supplier Selleck Chemical LLC Supplier No. S45781G
Tyloxapol (Triton WR1339) is a nonionic liquid polymer of the alkyl aryl polyether alcohol type used as a surfactant to aid liquefaction removal of mucopurulent and bronchopulmonary secretions It also blocks plasma lipolytic activity and thus the breakdown of triglyceride-rich lipoproteins Tyloxapol can be used to induce animal models of Hyperlipidemia Atherosclerosis