Learn More
Medchemexpress LLC Tylvalosin-d9 (Acetylisovaleryltylosin-d9) | 500 UG
SDP
Catalog No. 5000338923
Encompass_Preferred
Tylvalosin-d9 is the deuterium-labeled form of Tylvalosin, a third-generation macrolide with anti-inflammatory properties. It reduces levels of IL-8, IL-6, IL-1β, PGE2, TNF-α, and NO, and decreases inflammatory cell recruitment and activation in a mouse acute lung injury model. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Reduces levels of inflammatory mediators
  • Decreases inflammatory cell recruitment and activation
  • Functions as a tracer for quantitative analysis
  • Functions as an internal standard for techniques like NMR, GC-MS, or LC-MS
Catalog No. 50-003-38923 Supplier Medchemexpress LLC Supplier No. HY128423AS500UG
Tylvalosin-d9 is the deuterium-labeled form of Tylvalosin, a third-generation macrolide with anti-inflammatory properties. It reduces levels of IL-8, IL-6, IL-1β, PGE2, TNF-α, and NO, and decreases inflammatory cell recruitment and activation in a mouse acute lung injury model. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Reduces levels of inflammatory mediators
  • Decreases inflammatory cell recruitment and activation
  • Functions as a tracer for quantitative analysis
  • Functions as an internal standard for techniques like NMR, GC-MS, or LC-MS