Learn More
TARGETMOL CHEMICALS INC Tylvalosin tartrate 1G
SDP
Catalog No. 502877475
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Tylvalosin tartrate (Acetylisovaleryltylosin Tartrate) is a macrolide antibiotic that can against Gram-positive bacteria. Purity 98.01%
Catalog No. 50-287-7475 Supplier TARGETMOL CHEMICALS INC Supplier No. T222281G
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Tylvalosin tartrate (Acetylisovaleryltylosin Tartrate) is a macrolide antibiotic that can against Gram-positive bacteria. Purity 98.01%